Learn More
Lkt Laboratories CAS 51-55-8, ≥98%, mAChR antagonist found in Solanaceae used to initiate mydriasis. It decreases thermal pain, inhibits histamine-induced increases in thromboxane A2, and acts as a positive inotrope.
SDP
Supplier:  Lkt Laboratories A7868
Encompass_Preferred
Atropine is a tropane alkaloid found in plants in the Solanaceae family. Atropine is an anticholinergic compound that also exhibits analgesic and antinociceptive activities. Atropine is a competitive inhibitor of muscarinic acetylcholine receptors (mAChRs) that is commonly used for ophthalmic treatments such as initiating mydriasis. Atropine displays cardiostimulatory/positive inotropic activity as well, increasing heart rate by inhibiting vagal nerve activity. In animal models of thermal pain, atropine exhibits analgesic activity; in other animal models, atropine inhibits histamine-induced increases in thromboxane A2 (TxA2) levels.
Catalog No. NC2539822