Learn More
Medchemexpress LLC Atropine-d5 | 5908-99-6 | ≥99.0% | 1 MG
SDP
Catalog No. 5000407129
Encompass_Preferred
Atropine-d5 is a deuterium-labeled version of Atropine, which is a broad-spectrum, competitive antagonist of the muscarinic acetylcholine receptor (mAChR). This compound is valuable as a tracer and an internal standard for quantitative analysis, leveraging the influence of deuteration on pharmacokinetic and metabolic profiles in drug development.
  • Deuterium-labeled version of atropine.
  • Acts as a broad-spectrum, competitive antagonist of the muscarinic acetylcholine receptor.
  • Can be used as a tracer for quantitative analysis.
  • Serves as an internal standard for techniques like NMR, GC-MS, or LC-MS.
  • Useful in drug development for pharmacokinetic and metabolic profiling.
Catalog No. 50-004-07129 Supplier Medchemexpress LLC Supplier No. HYB0394S1MG
Add to Cart
Add to Cart
Atropine-d5 is a deuterium-labeled version of Atropine, which is a broad-spectrum, competitive antagonist of the muscarinic acetylcholine receptor (mAChR). This compound is valuable as a tracer and an internal standard for quantitative analysis, leveraging the influence of deuteration on pharmacokinetic and metabolic profiles in drug development.
  • Deuterium-labeled version of atropine.
  • Acts as a broad-spectrum, competitive antagonist of the muscarinic acetylcholine receptor.
  • Can be used as a tracer for quantitative analysis.
  • Serves as an internal standard for techniques like NMR, GC-MS, or LC-MS.
  • Useful in drug development for pharmacokinetic and metabolic profiling.