Learn More
Medchemexpress LLC HY-112076 50mg Medchemexpress, Atropine methyl bromide CAS:2870-71-5 Purity:>98%
SDP
Catalog No. 501969016
Encompass_Preferred
Medchemexpress, HY-112076 50mg Atropine methyl bromide CAS:2870-71-5 Atropine methyl bromide, a muscarinic receptor (mAChR) antagonist, is a quaternary ammonium salt of atropine and a mydriatic for dilation of the pupil during ophthalmic examination. It is introduced for relieving pyloric spasm in infants for its highly polar nature. It penetrates less readily into the central nervous system than atropine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9016 Supplier Medchemexpress LLC Supplier No. HY11207650MG
Medchemexpress, HY-112076 50mg Atropine methyl bromide CAS:2870-71-5 Atropine methyl bromide, a muscarinic receptor (mAChR) antagonist, is a quaternary ammonium salt of atropine and a mydriatic for dilation of the pupil during ophthalmic examination. It is introduced for relieving pyloric spasm in infants for its highly polar nature. It penetrates less readily into the central nervous system than atropine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.