Learn More
Medchemexpress LLC HY-B0394 100mg Medchemexpress, Atropine (sulfate monohydrate) CAS:5908-99-6 Purity:>98%
SDP
Catalog No. 502082516
Encompass_Preferred
Medchemexpress, HY-B0394 100mg Atropine (sulfate monohydrate) CAS:5908-99-6 Atropine (Tropine tropate) sulfate monohydrate is a broad-spectrum and competitive muscarinic acetylcholine receptor (mAChR) antagonist with anti-myopia effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2516 Supplier Medchemexpress LLC Supplier No. HYB0394100MG
Medchemexpress, HY-B0394 100mg Atropine (sulfate monohydrate) CAS:5908-99-6 Atropine (Tropine tropate) sulfate monohydrate is a broad-spectrum and competitive muscarinic acetylcholine receptor (mAChR) antagonist with anti-myopia effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.