Learn More
Medchemexpress LLC Atropine sulfate monohydrate | 5908-99-6 | 99.9% | 50 MG
SDP
Catalog No. 5000407290
Encompass_Preferred
Atropine sulfate monohydrate is a competitive muscarinic acetylcholine receptor (mAChR) antagonist. With IC50 values of 0.39 nM for Human mAChR M4 and 0.71 nM for Chicken mAChR M4, it inhibits ACh-induced relaxations in human pulmonary veins. This compound can be used for research related to anti-myopia and bradycardia.
  • Acts as a competitive muscarinic acetylcholine receptor (mAChR) antagonist.
  • Inhibits ACh-induced relaxations in human pulmonary veins.
  • Suitable for research on anti-myopia and bradycardia.
Catalog No. 50-004-07290 Supplier Medchemexpress LLC Supplier No. HYB039450MG
Atropine sulfate monohydrate is a competitive muscarinic acetylcholine receptor (mAChR) antagonist. With IC50 values of 0.39 nM for Human mAChR M4 and 0.71 nM for Chicken mAChR M4, it inhibits ACh-induced relaxations in human pulmonary veins. This compound can be used for research related to anti-myopia and bradycardia.
  • Acts as a competitive muscarinic acetylcholine receptor (mAChR) antagonist.
  • Inhibits ACh-induced relaxations in human pulmonary veins.
  • Suitable for research on anti-myopia and bradycardia.