Learn More
Medchemexpress LLC Atropine sulfate monohydrate | 5908-99-6 | MFCD00074815 | 99.9% | 1 ML
SDP
Catalog No. 5000407293
Encompass_Preferred
Atropine sulfate monohydrate is a competitive muscarinic acetylcholine receptor (mAChR) antagonist. It demonstrates high potency with IC50 values of 0.39 nM for Human mAChR M4 and 0.71 nM for Chicken mAChR M4. This compound is known to inhibit ACh-induced relaxations in human pulmonary veins.
  • Competitive muscarinic acetylcholine receptor (mAChR) antagonist.
  • Inhibits ACh-induced relaxations in human pulmonary veins.
  • Suitable for research on anti-myopia.
  • Suitable for research on bradycardia.
  • Available as a solid, white to off-white appearance.
Catalog No. 50-004-07293 Supplier Medchemexpress LLC Supplier No. HYB039410MM/1ML
Atropine sulfate monohydrate is a competitive muscarinic acetylcholine receptor (mAChR) antagonist. It demonstrates high potency with IC50 values of 0.39 nM for Human mAChR M4 and 0.71 nM for Chicken mAChR M4. This compound is known to inhibit ACh-induced relaxations in human pulmonary veins.
  • Competitive muscarinic acetylcholine receptor (mAChR) antagonist.
  • Inhibits ACh-induced relaxations in human pulmonary veins.
  • Suitable for research on anti-myopia.
  • Suitable for research on bradycardia.
  • Available as a solid, white to off-white appearance.