Learn More
Sigma Aldrich Fine Chemicals Biosciences Atropine sulfate salt mono25G
Catalog No. 502903782
Encompass
Atropine is isolated from Atropa belladonna Datura stramonium and Mandragora officinarum. It is a tertiary amine which acts as a cholinergic receptor antagonist.
Catalog No. 50-290-3782 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A025725G
Atropine is isolated from Atropa belladonna Datura stramonium and Mandragora officinarum. It is a tertiary amine which acts as a cholinergic receptor antagonist.