Learn More
Sigma Aldrich Fine Chemicals Biosciences Atropine sulfate salt mono5G
Catalog No. 502897989
Encompass
Atropine is isolated from Atropa belladonna Datura stramonium and Mandragora officinarum. It is a tertiary amine which acts as a cholinergic receptor antagonist.
Catalog No. 50-289-7989 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A02575G
Atropine is isolated from Atropa belladonna Datura stramonium and Mandragora officinarum. It is a tertiary amine which acts as a cholinergic receptor antagonist.