Learn More
Medchemexpress LLC Atrovastatin-PEG3-FITC | 1440755-31-6 | 98.2% | 1150.31 | C64H68FN5O12S | 5 MG
SDP
Catalog No. 5000222077
Encompass_Preferred
Atrovastatin-PEG3-FITC is a fluorescently labeled atorvastatin derivative used as a ligand/tracer to study KRAS-PDEδ interactions. It is applied in fluorescence anisotropy and related binding assays to characterize protein-ligand interactions and support biochemical screening and biophysical analysis.
  • Fluorescent FITC label suitable for anisotropy and fluorescence assays.
  • Acts as a ligand for studying KRAS-PDEδ interactions.
  • High purity (98.17%).
  • Molecular weight 1150.31; formula C64H68FN5O12S.
  • DMSO soluble at 100 mg/mL; ultrasonic assistance and freshly opened DMSO recommended.
  • Available in small research pack sizes (for example, 5 mg).
Catalog No. 50-002-22077 Supplier Medchemexpress LLC Supplier No. HY1349775MG
Atrovastatin-PEG3-FITC is a fluorescently labeled atorvastatin derivative used as a ligand/tracer to study KRAS-PDEδ interactions. It is applied in fluorescence anisotropy and related binding assays to characterize protein-ligand interactions and support biochemical screening and biophysical analysis.
  • Fluorescent FITC label suitable for anisotropy and fluorescence assays.
  • Acts as a ligand for studying KRAS-PDEδ interactions.
  • High purity (98.17%).
  • Molecular weight 1150.31; formula C64H68FN5O12S.
  • DMSO soluble at 100 mg/mL; ultrasonic assistance and freshly opened DMSO recommended.
  • Available in small research pack sizes (for example, 5 mg).