Learn More
Cayman Chemical AtrvastatIn lacton 10mg
SDP
Catalog No. 502983048
Encompass_Preferred
An active metabolite of atorvastatin; inhibits HMG-CoA reductase (IC50 = 0.007 µM for the rat liver enzyme); inhibits CYP2C9.1, CYP2C9.3, and P-gp (IC50s = 16.8, 5.62, and 3.1-5.2 µM, respectively)
Catalog No. 50-298-3048 Supplier Cayman Chemical Supplier No. 2095110 MG
An active metabolite of atorvastatin; inhibits HMG-CoA reductase (IC50 = 0.007 µM for the rat liver enzyme); inhibits CYP2C9.1, CYP2C9.3, and P-gp (IC50s = 16.8, 5.62, and 3.1-5.2 µM, respectively)