Learn More
Diagnostic Biosystems ATRX ATRX (D-5)
Catalog No. 502407630
Encompass_Preferred
ATRX is a member of the Snf2 family of helicase/ATPases, which contribute to the remodeling of the nucelosome structure in an ATPdependent manner and facilitate the initiation of transcription and replication. Central nervous system (CNS): loss of ATRX expression in 45% of anaplastic astrocytomas, 27% of anaplastic oligoastrocytomas, 10% of anaplastic oligodendrogliomas. Genetic signatures, including ATRX, may refine classification of gliomas
Catalog No. 50-240-7630 Supplier Diagnostic Biosystems Supplier No. MOB60005
ATRX is a member of the Snf2 family of helicase/ATPases, which contribute to the remodeling of the nucelosome structure in an ATPdependent manner and facilitate the initiation of transcription and replication. Central nervous system (CNS): loss of ATRX expression in 45% of anaplastic astrocytomas, 27% of anaplastic oligoastrocytomas, 10% of anaplastic oligodendrogliomas. Genetic signatures, including ATRX, may refine classification of gliomas