Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 546MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: 39f. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Raji, HeLa or A431 Cells. Pancreas, Adrenal, Ovary, Kidney, Prostate or Stomach. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human ATRX protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 280kDa.Chromosome: Xq21.1. Gene Symbol: ATRX.
Catalog No. 50-188-5071