Learn More
OHAUS Attachable 2 Tier Platform
Catalog No. 01804429 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-429 Shaker Attachment
1 options
Catalog No. 01-804-429 Supplier OHAUS Supplier No. 83041265
2nd Tier Platform doubles your shaking capacity.
  • Easily attached and secured without any tools.
  • For use on the Mini-300 Orbital-Genies or any Genie Temp-Shaker models with Adhering Mat Platform or Flask Clamp Platform.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment
Model Attachable 2nd Tier Platform for Mini-300 or Temp Shaker models with Mat