Learn More
Cell Applications Inc ATTACHMENT FACTOR SOL 100ML
SDP
Catalog No. 50608886
Encompass
Promote attachment and spreading of endothelial cells. It is required for culturing microvascular endothelial cells 100 ml
Catalog No. 50-608-886 Supplier Cell Applications Inc Supplier No. 123100
Promote attachment and spreading of endothelial cells. It is required for culturing microvascular endothelial cells 100 ml