Learn More
Sigma Aldrich Fine Chemicals Biosciences Attachment factor Solution
Catalog No. 502901124
Encompass
Promotes attachment and spreading of endothelial cells. It is required for culturing microvascular endothelial cells.
Catalog No. 50-290-1124 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 123100
Promotes attachment and spreading of endothelial cells. It is required for culturing microvascular endothelial cells.