Learn More
Thomas Scientific Attest Biological Indicator For Steam, 100/pk
SDP
Catalog No. NC1997787
Encompass_Preferred
Indicator For Steam 270°F/ 132°C Vacuum Assisted or 250°F/ 121°C Gravity Sterilizers, 48 Hour Readout, Brown Cap, 100/pk.
Catalog No. NC1997787 Supplier Thomas Scientific Supplier No. 1251D05
Indicator For Steam 270°F/ 132°C Vacuum Assisted or 250°F/ 121°C Gravity Sterilizers, 48 Hour Readout, Brown Cap, 100/pk.