Learn More
MSC Nilfisk Wet/Dry Vacuum, Electric, 30 Liter Capacity, PTFE Filter
Catalog No. 502838192
Encompass
The ATTIX 33 wet/dry vacuum is engineered for professionals, delivering enhanced performance, productivity, and reduced maintenance. It features a 30 liter capacity and includes an InfiniClean™ washable main filter, making it suitable for various demanding applications.
  • InfiniClean system provides automatic filter cleaning for consistent high performance.
  • Durable, washable PTFE filter offers 99.9% filtration efficiency for fine dust.
  • Flat top for storing safety gear, accessories, or power tools.
  • Double-walled construction ensures durability.
  • Hooks and flexible rubber straps for easy storage and transportation.
  • Large wheels, locking casters, and optional handle enhance maneuverability.
  • Optional universal toolbox mount.
  • Auto tool start feature.
  • Onboard accessory storage.
Catalog No. 50-283-8192 Supplier MSC Supplier No. 38686333
The ATTIX 33 wet/dry vacuum is engineered for professionals, delivering enhanced performance, productivity, and reduced maintenance. It features a 30 liter capacity and includes an InfiniClean™ washable main filter, making it suitable for various demanding applications.
  • InfiniClean system provides automatic filter cleaning for consistent high performance.
  • Durable, washable PTFE filter offers 99.9% filtration efficiency for fine dust.
  • Flat top for storing safety gear, accessories, or power tools.
  • Double-walled construction ensures durability.
  • Hooks and flexible rubber straps for easy storage and transportation.
  • Large wheels, locking casters, and optional handle enhance maneuverability.
  • Optional universal toolbox mount.
  • Auto tool start feature.
  • Onboard accessory storage.