Learn More
Sigma Aldrich Fine Chemicals Biosciences Atto 488 azide1MG
Catalog No. 502901345
Encompass
Atto 488 is a superior fluorescence label with high molecular absorption (90.000) and quantum yield (0.80) as well as sufficient stokes shift. It is optimised for excitation with argon laser and is characterized by high photo stability.The azide modification is suitable for reactions with alkyne groups (Huisgen reaction - Click Chemistry).
Catalog No. 50-290-1345 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 727091MG
Atto 488 is a superior fluorescence label with high molecular absorption (90.000) and quantum yield (0.80) as well as sufficient stokes shift. It is optimised for excitation with argon laser and is characterized by high photo stability.The azide modification is suitable for reactions with alkyne groups (Huisgen reaction - Click Chemistry).