Learn More
Sigma Aldrich Fine Chemicals Biosciences Atto 488 Protein Labeling
Catalog No. 502908909
Encompass
This kit contains sufficient amounts of reactive dye buffers and protein purification sets for performing five labeling reactions (1 mg protein each) and for the subsequent purification of the labeled protein.
Catalog No. 50-290-8909 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 383711KT
This kit contains sufficient amounts of reactive dye buffers and protein purification sets for performing five labeling reactions (1 mg protein each) and for the subsequent purification of the labeled protein.