Learn More
Sigma Aldrich Fine Chemicals Biosciences Atto 488 suitable for fluo1MG
Catalog No. 502891559
Encompass
Atto 488 is a labeling dye with high molecular absorption (90,000) and quantum yield (0.80) as well as sufficient Stokes shift between excitation and emission maximum. It is optimized for excitation with an argon laser, and is characterized by high photostability.
Catalog No. 50-289-1559 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 410511MGF
Atto 488 is a labeling dye with high molecular absorption (90,000) and quantum yield (0.80) as well as sufficient Stokes shift between excitation and emission maximum. It is optimized for excitation with an argon laser, and is characterized by high photostability.