Learn More
Sigma Aldrich Fine Chemicals Biosciences Atto 490LS NHS ester suita1MG
Catalog No. 502900845
Encompass
Atto 490LS is a new fluorescent label featuring an extraordinary large Stokes-Shift of 165 nm. Thus the emission spectrum is almost completely separated from its absorption spectrum making the dye highly suitable for multiplexing experiments in particular in combination with Atto 488 and Atto 514. Atto 490LS is very hydrophillic and shows excellent water solubility. The dye exhibits a relatively high fluorescence quantum yield which is only slightly reduced after conjugation to biomolecules e.g. proteins even at high degrees of labeling (DOL). Atto 490LS is an anionic dye. After conjugation to a substrate the dye carries a net electrical charge of -1.find more information here
Catalog No. 50-290-0845 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 783621MG
Atto 490LS is a new fluorescent label featuring an extraordinary large Stokes-Shift of 165 nm. Thus the emission spectrum is almost completely separated from its absorption spectrum making the dye highly suitable for multiplexing experiments in particular in combination with Atto 488 and Atto 514. Atto 490LS is very hydrophillic and shows excellent water solubility. The dye exhibits a relatively high fluorescence quantum yield which is only slightly reduced after conjugation to biomolecules e.g. proteins even at high degrees of labeling (DOL). Atto 490LS is an anionic dye. After conjugation to a substrate the dye carries a net electrical charge of -1.find more information here