Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atto 565 NHS Ester BioReagent, Suitable for Fluorescence, 70% (Coupling to Amines), MilliporeSigma™ Supelco™

Catalog No. 111004448 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4448 1 mg
11-100-3400 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-4448 Supplier MilliporeSigma Supelco Supplier No. 724641MGF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescence of bioconjugates; employed in fluorescence resonance energy transfer, FRET, experiments.

  • Forms stable amide bonds with amino-containing compounds.
  • Provides strong absorption and high thermal and photostability.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type Fluorescent Dye
Content And Storage -20°C
Label Type Other Labels or Dyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.