Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atto 594 Maleimide BioReagent, Suitable for Fluorescence, 80% (Coupling Rate), MilliporeSigma™ Supelco™

Catalog No. 111004026 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4026 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4026 Supplier MilliporeSigma Supelco Supplier No. 087171MGF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Quantity 1 mg
Product Type Fluorescent Label
Content And Storage -20°C
Label Type Other Labels or Dyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.