Learn More
Sigma Aldrich Fine Chemicals Biosciences Atto 647N NHS ester BioRea1MG
Catalog No. 502901337
Encompass
Atto 647N is a superior red-emitting fluorescence dye with a strong absorption excellent fluorescence quantum yield (65%) high photostability excellent ozone resistance good solubility and very little triplet formation.
Catalog No. 50-290-1337 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 183731MGF
Atto 647N is a superior red-emitting fluorescence dye with a strong absorption excellent fluorescence quantum yield (65%) high photostability excellent ozone resistance good solubility and very little triplet formation.