Learn More
Sigma Aldrich Fine Chemicals Biosciences Atto MB2 NHS ester Methyle1MG
Catalog No. 502900059
Encompass
Atto MB2 NHS ester is a derivative of the well-known redox dye Methylene Blue. The dye can be reversibly reduced to the colorless leuko form. Upon oxidation (e.g. with oxygen) the dye recovers and the absorption is fully restored.The dye is suitable for labeling of DNA RNA proteins etc. In common with most Atto-labels the dye shows a high extinction coefficient.Atto MB2 NHS ester is moderately hydrophilic.find more information here
Catalog No. 50-290-0059 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 734991MG
Atto MB2 NHS ester is a derivative of the well-known redox dye Methylene Blue. The dye can be reversibly reduced to the colorless leuko form. Upon oxidation (e.g. with oxygen) the dye recovers and the absorption is fully restored.The dye is suitable for labeling of DNA RNA proteins etc. In common with most Atto-labels the dye shows a high extinction coefficient.Atto MB2 NHS ester is moderately hydrophilic.find more information here