Learn More
AFFINISEP USA LLC AttractSPE® Disks Tips RPS-H - Starter Kit
SDP
Catalog No. NC3429275
Encompass
AttractSPE® Disks Tips RPS-H - Starter Kit with spin adapters Tips for molecular biology purpose with sulfonic modified SDB membrane (RPS-H) - 200μL - T2 capacity - 25/pk + Spin adapters and collection tubes (25/pk). Starter kit offer valid for one box
Catalog No. NC3429275 Supplier AFFINISEP USA LLC Supplier No. KTADAPTTIPS RPSH. T2.2
AttractSPE® Disks Tips RPS-H - Starter Kit with spin adapters Tips for molecular biology purpose with sulfonic modified SDB membrane (RPS-H) - 200μL - T2 capacity - 25/pk + Spin adapters and collection tubes (25/pk). Starter kit offer valid for one box