Learn More
AFFINISEP USA LLC AttractSPE® Disks Tips RPS - Starter Kit
SDP
Catalog No. NC3429272
Encompass
AttractSPE® Disks Tips RPS - Starter Kit with spin adapters Tips for molecular biology purpose with sulfonic modified SDB membrane - 200μL - T2 capacity - 25/pk + Spin adapters and collection tubes (25/pk). Starter kit offer valid for one box
Catalog No. NC3429272 Supplier AFFINISEP USA LLC Supplier No. KTADAPTTIPS RPSM. T2.2
AttractSPE® Disks Tips RPS - Starter Kit with spin adapters Tips for molecular biology purpose with sulfonic modified SDB membrane - 200μL - T2 capacity - 25/pk + Spin adapters and collection tubes (25/pk). Starter kit offer valid for one box