Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Attribute Blocks
SCI_ED

Catalog No. S79363B
Change view
Click to view available options
Product Type:
Attribute Block Set
Giant set in Plastic storage tray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S79363B Giant set in Plastic storage tray
S79408 Attribute Block Set
S79363D -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S79363B Supplier Learning Resources, Inc Supplier No. LER 1272

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Order Info

Desk set sizes range from 10cm2 to 30cm2. Giant set sizes range from 25cm2 to 75cm2.

Specifications

Grade Level Early Childhood, Elementary, Intermediate - Middle School, High School
Grade Pre-K to 12
Format Hands-on Learning
Product Type Giant set in Plastic storage tray
Includes Giant set in Plastic storage tray
For Use With (Application) Teaches shapes, sorting, patterns, size and counting
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.