Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Attune™ CytPix™ Flow Cytometer, blue/violet 6

Catalog No. A51843
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51843 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A51843 Supplier Invitrogen™ Supplier No. A51843
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Attune CytPix Flow Cytometer combines bright-field images with high sensitivity and precision flow cytometry in a small footprint and is simple to use at any experience level.

The Attune CytPix Flow Cytometer combines bright-field images with high sensitivity and precision flow cytometry in a small footprint and is simple to use at any experience level. Designed and developed to work with the range of Attune family accessories, the Attune CytPix Flow Cytometer offers a variety of optical configuration options depending on your application needs so you can perform your multicolor analysis with the addition of images to aid in quality control of samples and data analysis.

Features of the Attune CytPix Flow Cytometer include:

  • Revolutionary speed and precision—acquire up to 10X data faster with acoustic and hydrodynamic focusing with electronic speeds up to 65,000 events/sec. High-speed imaging at a rate of 6,000 images/ sec with AI-driven image processing models increases analysis throughput.
  • Advanced analysis capabilities—offers image similarity calculations to quickly sort for like images, dimensionality reduction for visualizing population clustering, and 36 label-free morphology parameters to reduce complex gating schemes
  • Enhanced application tools—tailor image analysis with unlimited user-trained image processing models for comprehensive population statistics and quantitative measurements. Share trained models to improve standardization and encourage collaboration.
  • Versatility and support—efficiently process diverse samples with a clog-resistant mechanism. Includes maintenance and training support for optimal performance.
  • Intuitive and easy to learn—integrate images and image data seamlessly with existing protocols. Designed to empower flow analysis for any level user.

Unlike other image-enhanced flow cytometers, the Attune CytPix Flow Cytometer enables you to translate the images into data. The images are analyzed to provide distinct measurements such as particle count, size, shape, and surface complexity for each event. This enables you to gain unique insights, achieve higher accuracy, and feel confident in your analysis.

The Attune CytPix Flow Cytometer is ideal for anyone interested in high quality flow cytometry data combined with deeper, more insightful information on cell morphology, cell health, and overall sample quality.

Fast flow cytometry and imaging

The innovative combination of acoustic focusing technology coupled with traditional hydrodynamic focusing results in acquisition of flow data at speeds up to 10X faster than other flow cytometers. Additionally, acoustic focusing places cells closer to the focal plane for the high-speed camera, meaning there is no need to compromise on flow rate when acquiring traditional flow cytometry data sets. This helps reduce the time to obtain results.

Automated image processing and analysis

Thousands of high resolution bright-field images are rapidly analyzed by pre-trained machine learning models to interpret image characteristics into extended parameters. Rapid image processing is conducted in a user-defined queue where you can adjust processing speed and choose specific samples. You can correlate morphology with cell phenotypes in a quantitative way using traditional flow cytometry analysis without changing your protocol. The image-based measurements are displayed as parameters dot plots or histograms and can be plotted against standard flow cytometry parameters.

Fit for your lab today and in the future

The instrument's modular design provides the flexibility to help meet future capability needs with a variety of upgrade options. Purchase what you need now and easily upgrade later. The small footprint of the instrument requires minimal bench space and even allows for use in a cell culture biosafety hood.

  • Flexible—2 to 4 lasers, 14 colors, and 16 parameters
  • Field upgradeable—choose to add additional lasers when you decide
  • Customizable band pass filters for expanded fluorochrome choices

Tune into your data

The Attune CytPix Flow Cytometer can be used to easily analyze most samples, including cardiomyocytes. The system is designed with a clog-resistant syringe-driven system to push the sample with a force of up to 75 psi.

  • Combined imaging and flow cytometry data without the need to compromise existing protocols
  • Flexibility in sample concentrations and sample types
  • Elimination of time-consuming concentration steps with No-Wash, No-Lyse Filter Kit
  • Analysis of dilute samples, including CSF, stem cells, mouse bone marrow, mouse blood, thin needle aspirates, and other rare event sample types
  • Acquisition of a high volume of samples in minimal time
  • Quantify physical characteristics of thousands of leukocyte phenotypes with automated image analysis

Superior service and support

Included with the Attune CytPix Flow Cytometer is a planned maintenance visit during the warranty period to maximize instrument uptime. Also included with the Attune CytPix Flow Cytometer is a basic training with a member of our experienced flow cytometry support team to cover use and maintenance of the system.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Attune CytPix Flow Cytometer
• Minitower Windows 10 64-bit computer with Attune Cytometric Software
• Power cord with adapters
• USB cable
• Attune Shutdown Solution
• Attune Wash Solution
• Attune Focusing Fluid
• Attune NxT Flow Cell Cleaning Solution
• Attune Performance Tracking Beads
• Quick reference card

Format Tube(s), 384-well plate, 96-well plate
Laser Features Blue Excitation Laser (488 nm), Violet Excitation Laser (405 nm)
Frequency 50/60 Hz
Product Line Attune
Shipping Condition Room Temperature
Voltage 100/240 V
Quantity 1 each
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only, Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.