Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Attune™ CytPix™ Violet 6 Conversion Kit, for yellow laser

Catalog No. A50929
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50929 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A50929 Supplier Invitrogen™ Supplier No. A50929
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet. A conversion kit is required for each original, and for any upgraded lasers of the instrument.

  • Designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet
  • A conversion kit is required for each original, and for any upgraded lasers of the instrument
  • Conversion is carried out by a field service engineer
TRUSTED_SUSTAINABILITY

Specifications

Shipping Condition RT
Regulatory Status RUO
For Use With (Equipment) Attune CytPix Flow Cytometer
Product Line DJM
For Use With (Application) Flow Cytometry
Includes Filters, dichroics
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.