Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Attune™ NxT and CytPix Wash Bottle

Catalog No. 100022151
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-002-2151 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-002-2151 Supplier Thermo Scientific™ Supplier No. 100022151
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with the Attune™ NxT Acoustic Focusing Cytometer.

  • 175 mL replacement bottle for wash solution
TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Content And Storage Store at room temperature.
Quantity Each
Capacity 175 mL
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Wash Bottle

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.