Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Attune™ NxT and CytPix Waste Bottle

Catalog No. 502723661
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-272-3661 1 bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-272-3661 Supplier Invitrogen™ Supplier No. 100085928CST
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This is a 1.9-L replacement bottle for the waste tank on the Attune NxT Acoustic Focusing Cytometer.

  • This is a 1.9-L replacement bottle for the waste tank on the Attune NxT Acoustic Focusing Cytometer.
  • The bottle is designed for easy user replacement and with more robust fittings to prevent breakage.
  • This bottle replaces discontinued Cat. No. 100022156.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 bottle
For Use With (Equipment) Attune NxT
Content And Storage 1 Bottle
Product Line FPL
Shipping Condition Room temperature
Includes Waste Bottle
Product Type Flow Cytometer Accessory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.