Learn More
Medchemexpress LLC Atuliflapon (AZD5718) | 2041075-86-7 | 99.0% | 10 MG
SDP
Catalog No. 5000230504
Encompass_Preferred
Atuliflapon is an orally active FLAP (5-lipoxygenase activating protein) inhibitor used as a research tool to modulate leukotriene production and study inflammatory and cardiovascular biology.
  • Oral FLAP inhibitor with low-nanomolar potency in enzyme/binding assays.
  • Inhibits LTB4 production in human whole blood (IC50 ≈ 39 nM); binding Kd ≈ 4.4 nM in assay reports.
  • Chemical formula C24H26N6O3 and molecular weight 446.50 g/mol.
  • High purity (≈99.0%) suitable for in vitro studies.
  • Available as solids and as DMSO solutions (examples: 10 mg solid; 10 mM, 1 mL in DMSO).
Catalog No. 50-002-30504 Supplier Medchemexpress LLC Supplier No. HY12290810MG
Atuliflapon is an orally active FLAP (5-lipoxygenase activating protein) inhibitor used as a research tool to modulate leukotriene production and study inflammatory and cardiovascular biology.
  • Oral FLAP inhibitor with low-nanomolar potency in enzyme/binding assays.
  • Inhibits LTB4 production in human whole blood (IC50 ≈ 39 nM); binding Kd ≈ 4.4 nM in assay reports.
  • Chemical formula C24H26N6O3 and molecular weight 446.50 g/mol.
  • High purity (≈99.0%) suitable for in vitro studies.
  • Available as solids and as DMSO solutions (examples: 10 mg solid; 10 mM, 1 mL in DMSO).