Learn More
Medchemexpress LLC Atuliflapon (AZD5718) | 2041075-86-7 | 99.0% | 100 MG
SDP
Catalog No. 5000230680
Encompass_Preferred
Atuliflapon (AZD5718) is a small-molecule, orally active inhibitor of 5-lipoxygenase activating protein (FLAP) investigated as a research compound for cardiovascular indications such as coronary artery disease. It potently suppresses leukotriene production and has documented preclinical and clinical pharmacology data.
  • Oral FLAP inhibitor with potent activity (reported FLAP IC50 ≈ 2 nM).
  • Reduces leukotriene production in human whole blood.
  • Reported molecular weight 446.50 g·mol⁻¹ and formula C24H26N6O3.
  • High purity (about 99.0%), white to off-white solid.
  • Soluble in DMSO (≈125 mg/mL); sonication may be required for full dissolution.
  • Powder storage: -20°C for long-term stability; in solvent, store at -80°C when possible.
Catalog No. 50-002-30680 Supplier Medchemexpress LLC Supplier No. HY122908100MG
Atuliflapon (AZD5718) is a small-molecule, orally active inhibitor of 5-lipoxygenase activating protein (FLAP) investigated as a research compound for cardiovascular indications such as coronary artery disease. It potently suppresses leukotriene production and has documented preclinical and clinical pharmacology data.
  • Oral FLAP inhibitor with potent activity (reported FLAP IC50 ≈ 2 nM).
  • Reduces leukotriene production in human whole blood.
  • Reported molecular weight 446.50 g·mol⁻¹ and formula C24H26N6O3.
  • High purity (about 99.0%), white to off-white solid.
  • Soluble in DMSO (≈125 mg/mL); sonication may be required for full dissolution.
  • Powder storage: -20°C for long-term stability; in solvent, store at -80°C when possible.