Learn More
Medchemexpress LLC Atuliflapon (AZD5718) | 2041075-86-7 | 99.0% | 25 MG
SDP
Catalog No. 5000230539
Encompass_Preferred
Atuliflapon (AZD5718) is a small-molecule, orally active inhibitor of 5-lipoxygenase activating protein (FLAP) used in preclinical and clinical research to modulate leukotriene biosynthesis and investigate cardiovascular inflammation.
  • Inhibits FLAP with low-nanomolar potency in clinical studies.
  • Demonstrates target engagement via reduced LTB4 and LTE4 biomarkers.
  • Suitable for in vitro human and translational studies; species-dependent activity limits some animal models.
  • Provided as a solid powder with high purity for research use.
Catalog No. 50-002-30539 Supplier Medchemexpress LLC Supplier No. HY12290825MG
Atuliflapon (AZD5718) is a small-molecule, orally active inhibitor of 5-lipoxygenase activating protein (FLAP) used in preclinical and clinical research to modulate leukotriene biosynthesis and investigate cardiovascular inflammation.
  • Inhibits FLAP with low-nanomolar potency in clinical studies.
  • Demonstrates target engagement via reduced LTB4 and LTE4 biomarkers.
  • Suitable for in vitro human and translational studies; species-dependent activity limits some animal models.
  • Provided as a solid powder with high purity for research use.