Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Atuliflapon (AZD5718) | 2041075-86-7 | 99.0% | 5 MG
SDP

Catalog No. 5000231966
Encompass_Preferred

Atuliflapon (AZD5718) is a potent, orally active small-molecule inhibitor of FLAP (5-lipoxygenase activating protein) supplied as a solid research compound. It is reported to have an IC50 of 2 nM and is provided with high purity and defined storage conditions for preclinical and in vitro studies.

  • Potent FLAP inhibitor (IC50 ~2 nM)
  • White to off-white solid powder form
  • Molecular formula C24H26N6O3, molecular weight 446.50 g/mol
  • Reported purity 98.97%
  • Storage: powder -20°C (long term) or 4°C (short term)
  • Intended for research use only

Catalog No. 50-002-31966 Supplier Medchemexpress LLC Supplier No. HY1229085MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Atuliflapon (AZD5718) is a potent, orally active small-molecule inhibitor of FLAP (5-lipoxygenase activating protein) supplied as a solid research compound. It is reported to have an IC50 of 2 nM and is provided with high purity and defined storage conditions for preclinical and in vitro studies.

  • Potent FLAP inhibitor (IC50 ~2 nM)
  • White to off-white solid powder form
  • Molecular formula C24H26N6O3, molecular weight 446.50 g/mol
  • Reported purity 98.97%
  • Storage: powder -20°C (long term) or 4°C (short term)
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.