Learn More
Medchemexpress LLC Atuliflapon (AZD5718) | 2041075-86-7 | 99.0% | 50 MG
SDP
Catalog No. 5000230262
Encompass_Preferred
Atuliflapon (AZD5718) is a reversible inhibitor of 5-lipoxygenase activating protein (FLAP) developed for investigation of leukotriene-mediated inflammation and cardiovascular indications. It shows nanomolar potency against FLAP and has been evaluated in first-in-human and Phase 2a clinical studies. The compound is provided as a solid research sample for in vitro and translational research.
  • Reversible FLAP inhibitor with FLAP IC50 ≈ 2 nM.
  • Inhibits LTB4 production in human whole blood (IC50 ≈ 39 nM).
  • High purity research sample (98.97%).
  • Molecular weight 446.50 g/mol; chemical formula C24H26N6O3.
  • Suitable for in vitro and translational/clinical research; not for clinical use.
  • Pack size typically 50 mg solid sample.
Catalog No. 50-002-30262 Supplier Medchemexpress LLC Supplier No. HY12290850MG
Atuliflapon (AZD5718) is a reversible inhibitor of 5-lipoxygenase activating protein (FLAP) developed for investigation of leukotriene-mediated inflammation and cardiovascular indications. It shows nanomolar potency against FLAP and has been evaluated in first-in-human and Phase 2a clinical studies. The compound is provided as a solid research sample for in vitro and translational research.
  • Reversible FLAP inhibitor with FLAP IC50 ≈ 2 nM.
  • Inhibits LTB4 production in human whole blood (IC50 ≈ 39 nM).
  • High purity research sample (98.97%).
  • Molecular weight 446.50 g/mol; chemical formula C24H26N6O3.
  • Suitable for in vitro and translational/clinical research; not for clinical use.
  • Pack size typically 50 mg solid sample.