Learn More
Medchemexpress LLC HY-12871C 5mg Medchemexpress, Atuveciclib S-Enantiomer CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12871C5MG
Encompass_Preferred
Medchemexpress, HY-12871C 5mg Atuveciclib S-Enantiomer CAS: Atuveciclib S-Enantiomer (BAY-1143572 S-Enantiomer) is a potent and selective CDK9 inhibitor, which inhibits CDK9 / CycT1 with an IC 50 of 16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2634