Learn More
MSC EarthWay ATV Mount Broadcast Spreader, 80 lb, 12-Volt, Poly Hopper
Catalog No. 502837830
Encompass
The EarthWay M20 is a 12-Volt ATV mount broadcast spreader featuring an 80 lb rustproof poly hopper and a 12-volt motor. This spreader is designed for uniform distribution of various materials such as fertilizer, seed, and chemicals.
  • 80 lb rustproof poly hopper
  • Even spread 3-hole drop shut-off system for uniform coverage
  • 12-volt motor located inside the hopper, housed in a thermal-plastic case for performance and long life
  • Hopper cover included
  • Manufactured in the U.S.
  • Recommended operating speed of 3 mph
  • Includes a 1-year limited warranty
Catalog No. 50-283-7830 Supplier MSC Supplier No. 63821896
The EarthWay M20 is a 12-Volt ATV mount broadcast spreader featuring an 80 lb rustproof poly hopper and a 12-volt motor. This spreader is designed for uniform distribution of various materials such as fertilizer, seed, and chemicals.
  • 80 lb rustproof poly hopper
  • Even spread 3-hole drop shut-off system for uniform coverage
  • 12-volt motor located inside the hopper, housed in a thermal-plastic case for performance and long life
  • Hopper cover included
  • Manufactured in the U.S.
  • Recommended operating speed of 3 mph
  • Includes a 1-year limited warranty