Learn More
Medchemexpress LLC ATX-II TFA | 99.7% | 4934.62 | 500 UG
SDP
Catalog No. 5000424489
Encompass_Preferred
ATX-II TFA is a selective sodium channel modulator toxin that enhances late sodium current, prevents full sodium channel inactivation, and generates persistent current fractions. It has a pro-arrhythmic effect, slowing intrinsic heart rate and prolonging QT interval. This compound is suitable for research on atrial fibrillation, long QT syndrome, and long QT3 syndrome.
Catalog No. 50-004-24489 Supplier Medchemexpress LLC Supplier No. HYP1604A500UG
ATX-II TFA is a selective sodium channel modulator toxin that enhances late sodium current, prevents full sodium channel inactivation, and generates persistent current fractions. It has a pro-arrhythmic effect, slowing intrinsic heart rate and prolonging QT interval. This compound is suitable for research on atrial fibrillation, long QT syndrome, and long QT3 syndrome.