Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ATX-II TFA | 99.7% | 4934.62 (free acid) | 1 MG
SDP

Catalog No. 5000424371
Encompass_Preferred

ATX-II TFA is a selective sodium channel modulator toxin. It enhances late sodium current and prevents full sodium channel inactivation, leading to persistent current fractions. This can have pro-arrhythmic effects, such as slowing intrinsic heart rate and prolonging the QT interval. It is suitable for research on atrial fibrillation and long QT syndromes.

  • Selective sodium channel modulator toxin
  • Enhances late sodium current
  • Prevents full sodium channel inactivation
  • Generates persistent current fractions
  • Has a pro-arrhythmic effect
  • Slows intrinsic heart rate
  • Prolongs QT interval
  • Suitable for research on atrial fibrillation
  • Suitable for research on long QT syndrome
  • Purity of 99.7%
  • Molecular weight of 4934.62 (free acid)
  • Appearance as a solid
  • White to off-white color

Catalog No. 50-004-24371 Supplier Medchemexpress LLC Supplier No. HYP1604A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ATX-II TFA is a selective sodium channel modulator toxin. It enhances late sodium current and prevents full sodium channel inactivation, leading to persistent current fractions. This can have pro-arrhythmic effects, such as slowing intrinsic heart rate and prolonging the QT interval. It is suitable for research on atrial fibrillation and long QT syndromes.

  • Selective sodium channel modulator toxin
  • Enhances late sodium current
  • Prevents full sodium channel inactivation
  • Generates persistent current fractions
  • Has a pro-arrhythmic effect
  • Slows intrinsic heart rate
  • Prolongs QT interval
  • Suitable for research on atrial fibrillation
  • Suitable for research on long QT syndrome
  • Purity of 99.7%
  • Molecular weight of 4934.62 (free acid)
  • Appearance as a solid
  • White to off-white color

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.