Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ATX inhibitor 1 | 2225892-70-4 | 99.3% | 501.30 | 10 MM 1 ML
SDP

Catalog No. 5000343098
Encompass_Preferred

ATX inhibitor 1 is a potent ATX inhibitor, with IC50 values of 1.23 nM (FS-3) and 2.18 nM (bis-pNPP). It is supplied as a white to off-white solid.

  • Potent ATX inhibitor
  • Molecular weight of 501.30
  • Store at -20°C for up to 3 years as powder
  • Soluble in DMSO (250 mg/mL)

Catalog No. 50-003-43098 Supplier Medchemexpress LLC Supplier No. HY111410100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ATX inhibitor 1 is a potent ATX inhibitor, with IC50 values of 1.23 nM (FS-3) and 2.18 nM (bis-pNPP). It is supplied as a white to off-white solid.

  • Potent ATX inhibitor
  • Molecular weight of 501.30
  • Store at -20°C for up to 3 years as powder
  • Soluble in DMSO (250 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.