Learn More
Medchemexpress LLC ATX inhibitor 1 | 2225892-70-4 | 99.3% | 501.30 | 50 MG
SDP
Catalog No. 5000344143
Encompass_Preferred
ATX inhibitor 1 is a potent inhibitor of ATX (Autotaxin) with IC50 values of 1.23 nM (FS-3) and 2.18 nM (bis-pNPP). It is intended for research use only.
  • Potent ATX inhibitor.
  • IC50: 1.23 nM (FS-3) and 2.18 nM (bis-pNPP).
  • Appearance: Solid, white to off-white.
  • For research use only.
Catalog No. 50-003-44143 Supplier Medchemexpress LLC Supplier No. HY11141010MG
ATX inhibitor 1 is a potent inhibitor of ATX (Autotaxin) with IC50 values of 1.23 nM (FS-3) and 2.18 nM (bis-pNPP). It is intended for research use only.
  • Potent ATX inhibitor.
  • IC50: 1.23 nM (FS-3) and 2.18 nM (bis-pNPP).
  • Appearance: Solid, white to off-white.
  • For research use only.