Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Atx inhibitor 1 | 2225892-70-4 | 99.3% | 501.30 g/mol | C21H23Cl2N2O6P | 5 MG
SDP

Catalog No. 5000263595
Encompass_Preferred

ATX inhibitor 1 is a potent autotaxin (ATX) inhibitor with nanomolar activity (IC50 = 1.23 nM by FS-3 assay; 2.18 nM by bis-pNPP). It is supplied as a solid research compound for in vitro and preclinical research use only.

  • Potent autotaxin inhibitor with nanomolar IC50 values (1.23 nM FS-3; 2.18 nM bis-pNPP).
  • Molecular formula: C21H23Cl2N2O6P.
  • Molecular weight: 501.30 g/mol.
  • Purity: 99.3% by HPLC.
  • Physical form: solid powder.
  • Storage: powder -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months.
  • Available sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, or 10 mM in DMSO solutions.
  • For research use only; not for human therapeutic use.

Catalog No. 50-002-63595 Supplier Medchemexpress LLC Supplier No. HY1114105MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ATX inhibitor 1 is a potent autotaxin (ATX) inhibitor with nanomolar activity (IC50 = 1.23 nM by FS-3 assay; 2.18 nM by bis-pNPP). It is supplied as a solid research compound for in vitro and preclinical research use only.

  • Potent autotaxin inhibitor with nanomolar IC50 values (1.23 nM FS-3; 2.18 nM bis-pNPP).
  • Molecular formula: C21H23Cl2N2O6P.
  • Molecular weight: 501.30 g/mol.
  • Purity: 99.3% by HPLC.
  • Physical form: solid powder.
  • Storage: powder -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months.
  • Available sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, or 10 mM in DMSO solutions.
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.