Learn More
Medchemexpress LLC Atx inhibitor 11 | 2485779-27-7 | 99.4% | 585.65 g/mol | C32H35N5O6 | 5 MG
SDP
Catalog No. 5000222680
Encompass_Preferred
ATX inhibitor 11 is a research-grade small-molecule autotaxin (ATX) inhibitor with reported IC50 of 2.7 nM. Supplied as a high-purity powder for in vitro and in vivo studies, it has documented solubility in DMSO and defined cold-storage recommendations. The compound is used in preclinical fibrosis models, including lung fibrosis, to reduce α-SMA deposition.
  • High potency against autotaxin (IC50 2.7 nM).
  • High purity suitable for research applications.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Stable powder with defined cold-storage conditions.
  • Validated in preclinical fibrosis models to reduce α-SMA deposition.
Catalog No. 50-002-22680 Supplier Medchemexpress LLC Supplier No. HY1432375MG
ATX inhibitor 11 is a research-grade small-molecule autotaxin (ATX) inhibitor with reported IC50 of 2.7 nM. Supplied as a high-purity powder for in vitro and in vivo studies, it has documented solubility in DMSO and defined cold-storage recommendations. The compound is used in preclinical fibrosis models, including lung fibrosis, to reduce α-SMA deposition.
  • High potency against autotaxin (IC50 2.7 nM).
  • High purity suitable for research applications.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Stable powder with defined cold-storage conditions.
  • Validated in preclinical fibrosis models to reduce α-SMA deposition.