Learn More
TARGETMOL CHEMICALS INC ATX INHIBITOR 5 5MG
SDP
Catalog No. 502581726
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ATX inhibitor 5 is a potent and orally active autotaxin (ATX) inhibitor (IC50 15.3 nM) that reduces the level of CCl4-induced hepatic fibrosis and has anti-hepatic fibrosis effects. purity: 99%
Catalog No. 50-258-1726 Supplier TARGETMOL CHEMICALS INC Supplier No. T104095MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ATX inhibitor 5 is a potent and orally active autotaxin (ATX) inhibitor (IC50 15.3 nM) that reduces the level of CCl4-induced hepatic fibrosis and has anti-hepatic fibrosis effects. purity: 99%