Learn More
Aviva Systems Biology Corp ATXN10 Antibody - N-terminal region (ARP61891_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61891_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATXN10. It was validated on Western Blot. . Alternate names: E46L, FLJ37990, HUMEEP, SCA10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-500