Learn More
Aviva Systems Biology Corp ATXN2 Antibody - middle region (ARP38210_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38210_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATXN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATXN2. Alternate names: ATX2, FLJ46772, SCA2, TNRC13ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56565