Learn More
Aviva Systems Biology Corp ATXN3 Antibody - N-terminal region (ARP50507_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50507_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATXN3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ATXN3. Alternate names: AT3, ATX3, JOS, MJD, MJD1, SCA3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-514