Learn More
Medchemexpress LLC Au-15330 | 2380274-50-8 | 99.9% | C39H49N9O5S | 10MG
SDP
Catalog No. 5000260841
Encompass_Preferred
AU-15330 is a proteolysis-targeting chimera (PROTAC) that selectively degrades SWI/SNF ATPase subunits SMARCA2 and SMARCA4. It demonstrates potent antitumor activity in prostate cancer xenograft models and shows synergy with androgen receptor antagonists in preclinical studies.
  • Selective degradation of SMARCA2 and SMARCA4
  • Demonstrated antitumor activity in prostate cancer xenograft models
  • Synergizes with androgen receptor antagonists
  • High reported purity (≈99.9%) suitable for research use
  • Solid, off-white to yellow appearance for easy handling
  • Storage-stable when kept under recommended conditions
Catalog No. 50-002-60841 Supplier Medchemexpress LLC Supplier No. HY14538810MG
AU-15330 is a proteolysis-targeting chimera (PROTAC) that selectively degrades SWI/SNF ATPase subunits SMARCA2 and SMARCA4. It demonstrates potent antitumor activity in prostate cancer xenograft models and shows synergy with androgen receptor antagonists in preclinical studies.
  • Selective degradation of SMARCA2 and SMARCA4
  • Demonstrated antitumor activity in prostate cancer xenograft models
  • Synergizes with androgen receptor antagonists
  • High reported purity (≈99.9%) suitable for research use
  • Solid, off-white to yellow appearance for easy handling
  • Storage-stable when kept under recommended conditions